Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Thermo Scientific™
Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Generate an accurate turbidity reading quickly with this reliable Turbidity Meter and its accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
AC3CBRVial Cleaning Brushes
AC3V25Vials
Catalog number AC3CBR
Price (USD)
67.23
Online Exclusive
74.70
Save 7.47 (10%)
Each
Type:
Vial Cleaning Brushes
Price (USD)
67.23
Online Exclusive
74.70
Save 7.47 (10%)
Each
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Additional Accessory: 
  • Replacement Calibration Standards (see AC301S)
Specifications
DescriptionVial Cleaning Brushes, 5 Pack
For Use With (Equipment)AQUAfast AQ3010 Turbidity Meter
TypeVial Cleaning Brushes
Unit SizeEach