μPAC™ Neo Plus ESI-MS Accessories
Thermo Scientific™
μPAC™ Neo Plus ESI-MS Accessories
Use μPAC™ ESI-MS Accessories for fast, reproducible, and low dispersion grounded connection between Thermo Scientific™ μPAC™ Neo Plus HPLC columns and Thermo Scientific™ Nanospray Flex™ and EASY-Spray™ ion sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeLength (Metric)Diameter (Metric) Inner
ACC-VSP1006Voltage Spacer60 mm10 μm
ACC-VSP2006Voltage Spacer60 mm20 μm
ACC-VSP2050Voltage Spacer500 mm20 μm
ACC-NANOFIC10Interface60 mm10 μm
ACC-CAPFIC20Interface60 mm20 μm
ACC-GRNDCGrounding CableN/AN/A
Catalog number ACC-VSP1006
Price (USD)
750.00
Each
Type:
Voltage Spacer
Length (Metric):
60 mm
Diameter (Metric) Inner:
10 μm
Price (USD)
750.00
Each
Use Thermo Scientific™ μPAC™ ESI-MS Accessories to facilitate reproducible, low dispersion, and grounded connections between μPAC Neo Plus HPLC columns and low-flow mass spectrometer ion sources. μPAC Neo Plus voltage spacers are available for both EASY-Spray and Nanospray Flex ion sources. All configurations require a grounding cable.
μPAC Neo Plus voltage spacers
The following voltage spacers are available to support all µPAC Neo plus columns and multiple column oven configurations on EASY-Spray and NanoSpray Flex ion sources:
  • ACC-VSP1006 - 10 µm I.D. x 60 mm for use with flow rates < 1 µL/min and columns housed within an external column oven
  • ACC-VSP2006 - 20 µm I.D. x 60 mm for use with flow rates > 1 µL/min and columns housed within an external column oven
  • ACC-VSP2050 - 20 µm I.D. x 500 mm for use with flow rates < 3 µL/min and columns housed within the Vanquish Neo column compartment
μPAC Neo Plus Flex Ion Connect
The following Flex Ion Connect interfaces are available to support use of µPAC Neo Plus columns, the Nanospray Flex ion source, and any 360 µm O.D. emitter:
  • ACC-NANOFIC10 - 10 µm I.D. x 60 mm for use with flow rates < 1 µL/min and columns housed within an external column oven
  • ACC-CAPFIC20 - 20 µm I.D. x 60 mm for use with flow rates > 1 µL/min and columns housed within an external column oven
Specifications
Diameter (Metric) Inner10 μm
Length (Metric)60 mm
For Use With (Equipment)Low-flow ion sources, µPAC Neo Plus 50 and 110 cm columns
TypeVoltage Spacer
Unit SizeEach