Swine Anti-Goat Ig's, HRP

Catalog number: ACI0404

Invitrogen™  Related applications: Cellular Imaging | Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Swine whole molecule anti-goat immunoglobulin, not human Ig Adsorbed

Format/Formulation: HRP Conjugate
Analyte Specific Reagent. Analytical and performance characteristics are not established.

Specifications

Reactivity: Goat
Target Isotype: IgG
Host Species: Swine
Antibody Fragment: Whole Antibody
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 0.7 mg⁄mL
Shipping Condition: Approved for shipment on Wet or Dry Ice
Validated Application: Immunofluorescence

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols