IL12 (p40/p70) Mouse anti-Human Monoclonal Antibody (clone C8.1)

Catalog number: AHC0122

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

IL-12 (C8.1) UNCONJ MS X HU; Unconjugated Monoclonal antibody specific to Human IL-12. This antibody is validated for use in Immunoassay (ELISA). Anti-IL-12 recognizes the expressed product of the IL12A gene.
Analyte Specific Reagent. Analytical and performance characteristics are not established.

Specifications

Reactivity: Human
Validated Application: ELISA, Neutralizing
Host Species: Mouse
Clonality: Monoclonal
Clone ID: C8.1
Target Gene: IL-12 (p40/p70)
Gene ID: 3592
Label or Dye: Unconjugated
Product Size: 500 µg
Protein Family: Cytokines & Receptors
Research Category: Immunology
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products