Kappa/Lambda Double Stain Kit

Catalog number: AHM4907

Invitrogen™  Related applications: Flow Cytometry | Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Two separate reagents are included in this double staining reagent system. The anti-light chain reagent is a mixture of affinity-isolated goat F(ab')2 anti-human kappa conjugated to fluorescein (FITC) and goat F(ab')2 anti-human lambda conjugated to R-Phy
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human
Target Isotype: Kappa Light Chain, Lambda Light Chain
Host Species: Goat
Antibody Fragment: F(ab')2
Label or Dye: 2 Color
Number of Reactions: 100 Tests
Product Size: 100 tests
Shipping Condition: Wet Ice
Validated Application: Flow Cytometry

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products