Ammonium dihydrogen arsenate, 98%
Ammonium dihydrogen arsenate, 98%
Ammonium dihydrogen arsenate, 98%
Alfa Aesar

Ammonium dihydrogen arsenate, 98%

CAS: 13462-93-6 | AsH6NO4 | 158.973 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF-031493-30
HSN 28419000
Price (INR)
241,468.00
Each
Quantity:
250 g
Request bulk or custom format
Price (INR)
241,468.00
Each
Chemical Identifiers
CAS13462-93-6
IUPAC Nameammonium dihydrogen arsorate
Molecular FormulaAsH6NO4
InChI KeyVTWJXWNBCACAAK-UHFFFAOYSA-N
SMILES[NH4+].O[As](O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only