Graphite foil, 0.13mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Graphite foil, 0.13mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Graphite foil, 0.13mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Graphite foil, 0.13mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Graphite foil, 0.13mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Alfa Aesar

Graphite foil, 0.13mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 100 cm
30 x 30 cm
30 x 500 cm
Catalog number ALF-043078-CJ
HSN 38011000
Price (INR)
4,941.00
Each
Add to cart
Quantity:
30 x 100 cm
Request bulk or custom format
Price (INR)
4,941.00
Each
Add to cart
Specifications
Chemical Name or MaterialGraphite
CAS7782-42-5
ColorGray to Black
Recommended StorageAmbient temperatures
Density2g/cm3 at 20°C
View more

Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only