Formamidine acetate, 99%
Formamidine acetate, 99%
Formamidine acetate, 99%
Alfa Aesar

Formamidine acetate, 99%

CAS: 3473-63-0 | C3H8N2O2 | 104.109 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF-A11158-22
HSN 29252990
Price (INR)
6,347.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
6,347.00
Each
Chemical Identifiers
CAS3473-63-0
IUPAC Nameacetic acid; methanimidamide
Molecular FormulaC3H8N2O2
InChI KeyXPOLVIIHTDKJRY-UHFFFAOYSA-N
SMILESNC=N.CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)≤0.75%
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

Solubility
Soluble in water and hot alcohol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. More convenient source of formamidine than the corresponding chloride in that it is non-hygroscopic and, being the salt of a weaker acid, can often be used in synthesis without the need to liberate the free base.
  2. Widely used in heterocyclic syntheses, for example of pyrimidine and imidazole derivatives.
  3. Maiden, T. M. M.; Swanson, S.; Procopiou, P. A.; Harrity, J. P. A Mild and Regioselective Route to Functionalized Quinazolines. Chem. Eur. J. 2015, 21 (41), 14342-14346.
  4. Cebrián, C.; Natali, M.; Villa, D.; Panigati, M.; Mauro, M.; D'Alfonso, G.; De Cola, L. Luminescent supramolecular soft nanostructures from amphiphilic dinuclear Re(I) complexes. Nanoscale 2015, 7, 12000-12009.