Diisopropyl ketone, 98%
Diisopropyl ketone, 98%
Diisopropyl ketone, 98%
Alfa Aesar

Diisopropyl ketone, 98%

CAS: 565-80-0 | C7H14O | 114.188 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF-A12875-36
HSN 29141990
Price (INR)
8,286.00
Each
Quantity:
500 g
Request bulk or custom format
Price (INR)
8,286.00
Each
Chemical Identifiers
CAS565-80-0
IUPAC Name2,4-dimethylpentan-3-one
Molecular FormulaC7H14O
InChI KeyHXVNBWAKAOHACI-UHFFFAOYSA-N
SMILESCC(C)C(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.3980-1.4020 @ 20?C
Assay (GC)≥97.5%
2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
RUO – Research Use Only

General References:

  1. Wang Y, et al. Equilibrium 2H/1H fractionations in organic molecules: I. Experimental calibration of ab initio calculations.Geochim. Cosmochim. Acta.,2009,73(23), 7060-7075.
  2. Frank C. Whitmore and R. S. George. Abnormal Grignard Reactions. X.1 Enolizing and Reducing Action of Grignard Reagents upon Diisopropyl Ketone.J. Am. Chem. Soc.,1942,64(6), 1239-1242.