RNase-free Tips, 1000 μL size
Invitrogen™

RNase-free Tips, 1000 μL size

Ambion Tips are RNase- and DNase-free and are provided in ten racks containing 96 tips each. They are compatible withRead more
Have Questions?
Catalog NumberQuantity
AM12660TS10 racks
Catalog number AM12660TS
Price (USD)
180.00
10 racks
Quantity:
10 racks
Price (USD)
180.00
10 racks
Ambion Tips are RNase- and DNase-free and are provided in ten racks containing 96 tips each. They are compatible with Eppendorf, Finnpipette, Oxford Benchmate, Pipetman, Proline, Rainin, Socorex, and other brands of pipettors. Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free.
For research use only. Not for use in diagnostic procedures.
Specifications
Product TypeRNase-free Tips
Quantity10 racks
Shipping ConditionRoom Temperature
FilteredNon-filtered
For Use With (Equipment)Universal Fit
Product LineAmbion
SterilitySterile
Volume (Metric)1000 μL
Unit Size10 racks
Contents & Storage
Includes 10 racks with 96 tips each.

Store at room temperature.