MEGAscript™ SP6 Transcription Kit, 40 Reactions - FAQs
MEGAscript™ SP6 Transcription Kit, 40 Reactions - FAQs
View additional product information for MEGAscript™ SP6 Transcription Kit - FAQs (AM1330)
1 product FAQs found
I am using the MEGAscript SP6 Transcription Kit for in vitro transcription. I ran my reaction product on a denaturing gel and got a smear. Can you offer some advice?
If the RNA appears degraded (e.g., smeared), remove residual RNase from the DNA template preparation before in vitro transcription. Do this by digesting the DNA prep with proteinase K (100-200 µg/mL) in the presence of 0.5% SDS for 30 min at 50°C, and follow this with phenol/chloroform extraction. The RNase Inhibitor that is present in the transcription reaction can only inactivate trace RNase contamination. Large amounts of RNase contamination will compromise the size and amount of transcription products.