mMESSAGE mMACHINE™ T7 Transcription Kit
Invitrogen™

mMESSAGE mMACHINE™ T7 Transcription Kit

mMESSAGE mMACHINE kits are designed for the in vitro synthesis of large amounts of capped RNA. Capped RNA mimics most eukaryotic mRNAs found in vivo, because it has a 7-methyl guanosine cap structure at the 5' end.
Have Questions?
Catalog NumberQuantity
AM134425 Reactions
Catalog number AM1344
Price (USD)
548.00
Each
Quantity:
25 Reactions
Price (USD)
548.00
Each

mMESSAGE mMACHINE kits are designed for the in vitro synthesis of large amounts of capped RNA. Capped RNA mimics most eukaryotic mRNAs found in vivo, because it has a 7-methyl guanosine cap structure at the 5' end. mMESSAGE mMACHINE kit reactions include cap analog [m7G(5')ppp(5')G] in an ultra high-yield transcription reaction. The cap analog is incorporated only as the first or 5' terminal G of the transcript because its structure precludes its incorporation at any other position in the RNA molecule.

How mMESSAGE mMACHINE kits work

mMESSAGE mMACHINE kits have a simplified reaction format in which all four ribonucleotides and cap analog are mixed in a single solution. The cap analog:GTP ratio of this solution is 4:1, which is optimal for maximizing both RNA yield and the proportion of capped transcripts. mMESSAGE mMACHINE kits are ideal for the routine synthesis of capped RNAs for oocyte microinjection, in vitro translation, transfection, and other applications. The high yields are achieved by optimizing reaction conditions for RNA synthesis in the presence of high nucleotide concentrations. In addition, the RNA synthesized is protected from degradation by any contaminating ribonucleases that may be present with RNase inhibitor—a component of the Enzyme Mix.

Using mMESSAGE mMACHINE Kits

In a 20 μL reaction during a two hour incubation, mMESSAGE mMACHINE high yield capped RNA transcription will yield large mass amounts of capped RNA. Up to 10–50 times the yield obtained with conventional in vitro transcription reactions. The ratio of cap analog to GTP has been optimized to allow the best compromise between yield (15–35 μg) and proportion of transcripts that are capped (80%).

mMESSAGE mMACHINE kits also contain a LiCl precipitation solution that is efficient for separating proteins and unincorporated nucleotides (including free cap analog) from the capped RNA, allowing an increased efficiency of translation. mMESSAGE mMACHINE kits are only optimized for use with the polymerases included in the kit. Using a different polymerase may result in low yields. The mMESSAGE mMACHINE kits contain all the buffers and reagents necessary for 25 transcription reactions. Using the control template supplied with the kits (Xenopus elongation factor 1α, pTRI Xef), each mMESSAGE mMACHINE reaction will yield approximately 20–30 μg of RNA using T3 or T7 RNA polymerase or about 15–25 μg RNA using SP6 RNA polymerase.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions25 Reactions
Product LineAmbion, mMESSAGE mMACHINE
Product TypeT7 In Vitro Transcription kit
PromoterT7
Quantity25 Reactions
Shipping ConditionDry Ice
Starting MaterialPCR Products, Linearized DNA plasmid
Yield20-30 μg
Final Product TypeSynthetic mRNA, mRNA
FormatKit
Sample TypemRNA, Synthetic mRNA
Unit SizeEach
Contents & Storage
T7 Enzyme Mix, 10X Reaction Buffer 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.

Frequently asked questions (FAQs)

I am using the mMESSAGE mMACHINE T3 Transcription Kit for in vitro transcription. I ran my reaction product on a denaturing gel and got a smear. Can you offer some advice?

If the RNA appears degraded (e.g. smeared), remove residual RNase from the DNA template preparation before in vitro transcription. Do this by digesting the DNA prep with proteinase K (100-200 µg/mL) in the presence of 0.5% SDS for30 min at 50°C, follow this with phenol/chloroform extraction. The RNase Inhibitor that is present in the transcription reaction, can only inactivate trace RNase contamination. Large amounts of RNase contamination will compromise the size and amount of transcription products.

What is the sequence for the pTri-Xef control template that comes with mMESSAGE mMACHINE T7 Transcription Kit (Cat. No. AM1344)?

The pTri-Xef control template that comes with mMESSAGE mMACHINE T7 Transcription Kit (Cat. No. AM1344) is a linearized TRIPLEscript DNA plasmid containing the 1.85 kb Xenopus elongation factor 1α gene under the transcriptional control of tandem SP6, T7, and T3 promoters (pTRI-Xef 1). While we cannot provide a full map of the linearized plasmid, the insert sequence with the three promotors is as follows:

ACGATTTAGGTGACACTATAGAATACACGGAATTAATACGACTCACTATAGGGAGACTCGAGAATTAACCCTCACTAAAGGGAGGTACCGCGGATGCATGAATTCCGCCGGTGAGTTCTAACTATCCACCGCCAAACATCTAATCCACAATGGGAAAGGAAAAGACTCACATCAACATCGTTGTCATTGGACACGTAGATTCTGGAAAGTCCACAACAACTGGACATCTGATCTACAAATGTGGTGGTATCGACAAGAGAACCATCGAAAAGTTCGAGAAGGAAGCTGCTGAGATGGGAAAAGGCTCCTTCAAGTATGCCTGGGTCTTGGACAAACTGAAGGCCGAGCGTGAACGTGGTATTACCATTGACATCTCCCTGTGGAAATTTGAGACCAGCAAATACTATGTCACTATCATTGATGCTCCTGGACACAGAGATTTCATCAAGAACATGATCACTGGTACTTCTCAGGCTGACTGTGCTGTCCTGATTGTTGCTGCTGGTGTTGGTGAATTTGAAGCTGGTATCTCAAAGAATGGACAAACCCGTGAGCATGCCCTCCTTGCCTACACTCTGGGTGTAAAACAGCTGATCGTTGGTATTAACAAGATGGATTCCACTGAACCCCCATACAGTCAGAAAAGATATGAGGAAATTGTAAAGGAAGTCAGCACATACATCAAGAAGATTGGTTACAACCCTGACACTGTTGCCTTTGTACCTATTTCTGGATGGAACGGTGACAACATGCTTGAGCCTAGCCCCAATATGCCATGGTTTAAGGGATGGAAAATCACCCGTAAAGAAGGATCTGGCAGCGGAACTACCCTGCTGGAAGCTCTTGACTGCATTCTGCCACCTAGTCGCCCAACTGATAAGCCTCTCCGTCTGCCTCTGCAGGATGTCTACAAAATTGGCGGTATTGGTACTGTACCAGTTGGTCGTGTGGAAACTGGTGTCATTAAACCAGGCATGGTGGTTACCTTTGCCCCTGTTAATGTAACAACTGAAGTGAAATCTGTTGAAATGCACCATGAAGCCCTTACTGAGGCCGTGCCCGGTGACAACGTCGGCTTTAACGTCAAGAACGTTTCCGTGAAGGACGTCCGTCGTGGCAACGTTGCTGGTGACAGCAAGATTGACCCACCAATGGAAGCTGGTTCTTTTACCGCACAGGTTATCATCCTGAATCACCCAGGCCAGATTGGTGCTGGATATGCCCCTGTGTTGGATTGCCACACTGCTCACATTGCTTGCAAGTTTGCTGAGCTAAAGGAGAAGATTGATCGCCGTTCTGGTAAGAAACTGGAAGACAATCCCAAGTTCCTGAAGTCTGGAGATGCTGCCATTGTTGACATGATCCCAGGAAAGCCAATGTGCGTGGAGAGCTTCTCAGACTACCCTCCTCTTGGTCGTTTTGCTGTCCGTGACATGAGGCAGACTGTTGCCCTAGGAGTCATCAAGGCAGTGGAAAAGAAGGCAGCAGGATCTGGCAAAGTCACAAAGTCTGCTCAGAAAGCAGCGAAAACCAAATGAATATTTCCAGAGTCTCTCACCTCAGTCATAACCAGTGGTGGAGGATTGGTCTCAGAACTCTTTCTATCAATTGGCCATCAGAGTTTAATAGTAAAAGACTGGTTAATGATTACAATGCATCGCAAGAGCTTCAGAAGGAAAAAAATGCTTGTGGACACATTTGTTTGTGGCAGTTTTAAGTTATTAGTGTTTTAAATCCAGTAATTTCTAAATGGAAGCAAATTGACCAAAATCTGTCACCAAATTTGAGACCATTAAAAAAAAGTTGAAAAGATAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAATAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAAACGGAATTTCGCGGAATTCGAGCTCGCCCGGGGATCCTCTAGAGTCGACCTGCAGCCCA

SP6 (ATTTAGGTGACACTATAGA) 1.92 kb
T7 (TAATACGACTCACTATAGG) 1.89 kb
T3 (AATTAACCCTCACTAAAGG) 1.86 kb

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.