MessageAmp™ II aRNA Amplification Kit, 20 Reactions - FAQs
MessageAmp™ II aRNA Amplification Kit, 20 Reactions - FAQs
View additional product information for MessageAmp™ II aRNA Amplification Kit - FAQs (AM1751)
1 product FAQs found
Can I use modified nucleotides with the MessageAmp II aRNA Amplification Kit?
Yes,modified nucleotides with the MessageAmp II aRNA Amplification Kit and instructions can be found in Section II.F., Page 17 of the manual (https://tools.thermofisher.com/content/sfs/manuals/cms_056141.pdf).