Amino Allyl MessageAmp™ II aRNA Amplification Kit, with Cy™3 dye
Catalog number: AM1795
Ambion™  Related applications: Microarray Analysis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
The Ambion® Amino Allyl MessageAmp™ aRNA Amplification Kit includes reagents for aRNA amplification with amimo allyl NTP incorporation. The kit includes sufficient reagents for 20 reactions. Cy™3 dye is included and packaged as individual tubes sufficient for one labeling reaction.
Improved First- and Second-strand cDNA Synthesis
Included in the kit is ArrayScript™ RT, a rationally engineered M-MLV reverse transcriptase, which produces equivalent or higher yields of full-length cDNA compared to other enzymes. The second-strand cDNA synthesis reaction has also been optimized to be compatible with the first-strand cDNA products generated with ArrayScript™ RT to allow maximal conversion of first-strand cDNA into full-length double-stranded cDNA templates.
Increased Labeling Efficiency of Your aRNA
The Amino Allyl MessageAmp™ aRNA Amplification Kit incorporates amino allyl NTP into the aRNA followed by the coupling of its reactive amino group to an NHS ester label (e.g., Cy™ or another dye). This strategy offers several advantages over the direct incorporation of labeled NTPs. Direct incorporation of labeled NTPs is inefficient and results in low yields and low specific activity aRNA and high costs. Unlike lableled NTPs, amino allyl–modified NTPs are incorporated almost as efficiently as unmodified NTPs and are much less expensive than the dye coupled NTPs.
For Research Use Only. Not for use in diagnostic procedures.
Product Size: 20 reactions
Labeling Method: Indirect Labeling
Includes Label or Dye: Yes
Reverse Transcriptase: ArrayScript™ Reverse Transcriptase
Sample Type (Specific): Total RNA, Poly(A+) RNA
Contents & storage
The Amino Allyl MessageAmp™ II aRNA Amplification Kit contains T7 Oligo(dT) Primer, ArrayScript™ RT, RNase Inhibitor, 10X First-Strand Buffer, dNTP Mix, 10X Second- Strand Buffer, DNA Polymerase, RNase H, T7 Enzyme Mix, T7 10X Reaction Buffer, T7 UTP Solution, aa-UTP Solution, ATP⁄CTP⁄GTP Mix, Second Round Primers, Coupling Buffer, DMSO, Hydroxylamine, Cy™3 dye, and Control RNA (1 mg⁄mL HeLa total RNA) which are all stored at –20°C. Wash Buffer, cDNA Binding Buffer, aRNA Binding Buffer, aRNA Filter Cartridges, aRNA Collection Tubes, cDNA Filter Cartridges, cDNA Elution Tubes, Nuclease-free Water, and detailed Instruction Manual are stored at room temperature.
Manuals & protocols