RNAqueous™-96 Automated Kit
Invitrogen™

RNAqueous™-96 Automated Kit

The Ambion™ RNAqueous™-96 Automated Kit is used for automated, phenol-free total RNA isolation from 100-500,000 cells or 0.1–1 mg tissueRead more
Have Questions?
Catalog NumberQuantity
AM1812384 Preps
Catalog number AM1812
Price (USD)
1,400.65
Online exclusive
1,804.00
Save 403.35
Each
Quantity:
384 Preps
Price (USD)
1,400.65
Online exclusive
1,804.00
Save 403.35
Each
The Ambion™ RNAqueous™-96 Automated Kit is used for automated, phenol-free total RNA isolation from 100-500,000 cells or 0.1–1 mg tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. The kit contains enough reagents for 384 (4 x 96) RNA purifications. The RNAqueous™-96 Automated Kit uses an RNA-binding glass-fiber filter to provide high, reproducible yields of intact RNA. An optional on-the-filter DNase treatment can be performed to ensure removal of genomic DNA for RT-PCR applications. Downloadable protocols for the use of this kit with specific liquid handling systems are available at our Automation Resource.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume105 μL
Final Product TypeTotal RNA
For Use With (Application)RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
For Use With (Equipment)MultiPROBE™ II HT, Biomek™ 2000, Liquid Handling Systems
High-throughput CompatibilityHigh-throughput Compatible
Quantity384 Preps
Shipping ConditionBox 1: Dry Ice
Box 2: Room Temperature
Starting Material AmountCells: ≤105
Tissue: 0.1–1 mg
Isolation TechnologyVacuum Filter Plate (Glass Fiber Filter)
Sample TypeCells, Tissue
Unit SizeEach
Contents & Storage
• 4 Filter Plates; store at room temperature
• 4 Collection Plates with lids; room temperature
• 4 Culture Plates; room temperature
• 1 Heat Plate with Lid; room temperature
• 100 mL Nuclease-free Water; room temperature
• 100 mL Elution Solution; room temperature
• 100 mL Lysis/Binding Solution; 4°C
• 100 mL Rebinding Mix; 4°C
• 2 x 175 mL Wash Solution (concentrated); 4°C
• 1.25 mL DNase 1; -20°C
• 12 mL DNase 1 Buffer; -20°C

Frequently asked questions (FAQs)

What are the differences in the RNAqueous kits you offer?

RNaqueous kits are based on a rapid, filter-based RNA isolation system that does not require phenol, chloroform, or other toxic organic chemicals. The RNAqueous Total RNA Isolation Kit is designed for sample sizes of 10-75 mg of tissue or 10^6 to 10^7 cells, whereas the RNAqueous-Midi Total RNA Isolation Kit is designed for tissue samples of up to 0.5 g or 10^9 cells. The RNAqueous-Micro Total RNA Isolation Kit is optimized for the purification of total RNA from micro-sized samples such as 10-500,000 cultured cells, as few as 10 microdissected cells, or up to 10 mg tissue. The RNAqueous-96 Total RNA Isolation Kit utilizes a 96-well plate format for high-throughput RNA isolation from 100 to 2 x 10^6 cells or 0.1 to 1.5 mg of tissue. The RNAqueous-4PCR Total RNA Isolation Kit is designed for RNA isolation from small tissue samples (0.5-75 mg) and small- to medium-sized cultured cell samples (approximately 100-10^7 cells).