This product will be discontinued on December 1, 2016. For superior performance, we recommend you switch to Platinum Taq DNA Polymerase High Fidelity
SuperTaq™ Plus Polymerase (cloned) 5 U/μL
Catalog number: AM2056
Applied Biosystems™  Related applications: PCR
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
This extended range Ambion® Taq Polymerase contains a proofreading activity that reduces the error rate of Taq Polymerase. It produces significantly higher yields of PCR products than ordinary Taq Polymerase, especially for fragments >1 kb, and can amplify up to 20 kb. It is supplied in one tube containing 250 U at a concentration of 5 U/ µL. It can be used in PCR and RT-PCR reactions. SuperTaq™ Plus is suitable as a direct replacement for ordinary Taq Polymerase in most applications.
Unit Definition:
One unit incorporates 10 nmol of deoxynucleotides into acid insoluble material in 30 min at 74°C. SuperTaq™ Plus is a trademark of, and is manufactured by Enzyme Technologies Ltd. and sold under licensing arrangements with F. Hoffman-La Roche, Ltd., Roche Molecular Systems, Inc. and the Perkin-Elmer Corporation. We are a distributor of Enzyme Technologies, Ltd., except in the following countries: Austria, Benelux, Denmark, Greece, Sweden, Switzerland, Taiwan, and the United Kingdom.
Contents & storage
SuperTaq Plus Polymerase, 10X Long PCR Buffer, 10X Long PCR Buffer without MgSO4, MgSO4, dNTP Mix. Store at –20°C.
Documents
Manuals & protocols