Ambion™ T7 RNA Polymerase, cloned, 200 U/μL
Invitrogen™

Ambion™ T7 RNA Polymerase, cloned, 200 U/μL

This cloned high-purity Ambion T7 RNA Polymerase has a high specificity for its promoter. The enzyme will transcribe large amountsRead more
Have Questions?
Catalog NumberQuantity
AM208530,000 units
Catalog number AM2085
Price (USD)
802.00
Each
Quantity:
30,000 units
Request bulk or custom format
Price (USD)
802.00
Each
This cloned high-purity Ambion T7 RNA Polymerase has a high specificity for its promoter. The enzyme will transcribe large amounts of RNA from DNA sequences downstream of the promoter and show no cross talk between promoters. One tube containing 30,000 U at a concentration of 200 U/μL is provided. Its high purity makes them ideal for synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA. The RNA Polymerase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity. Functionality of enzyme is tested in an in vitro transcription reaction under optimal and limiting UTP concentration.

Unit Definition:
One unit is the amount of T7 RNA polymerase required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity30,000 units
Shipping ConditionDry Ice
PolymeraseT7 RNA Polymerase
Product LineAmbion
Unit SizeEach
Contents & Storage
Store at -20°C.