TURBO DNase™ (2 U/μL), 5000 U - FAQs
TURBO DNase™ (2 U/μL), 5000 U - FAQs
View additional product information for TURBO DNase™ (2 U/μL) - FAQs (AM2239, AM2238)
15 product FAQs found
How do I remove DNase from an RNA sample?
DNase can be removed by the following methods: acid phenol:choloform extraction, lithium chloride precipitation, EDTA, and heat inactivation, or using our MegaClear Transcription Clean-Up Kit (Cat. No. AM1908).
Alternatively, our Invitrogen TURBO DNA-free Kit (Cat. No. AM1907) is supplied with a DNase inactivation reagent that can be used to remove the DNase, as well as divalent cations such as magnesium and calcium, which can catalyze RNA degradation when RNA is heated with the sample.
How can I inactivate DNase I?
Add of 1 µL of 25 mM EDTA solution to the reaction mixture in 10 µL reaction with 1 unit DNase I, Amplification Grade (or 1:1 molar ratio of Mg++ ions:EDTA) to chelate the Mg++ ions in the DNase I buffer. Heat for 10 min at 65 degrees C.
Note: It is vital that the EDTA be at least at 2 mM prior to heat-inactivation to avoid Mg-dependent RNA hydrolysis.
DNA-free and Turbo-free versions of DNase I can be inactivated with included DNase Inactivation Reagent.
Are your DNase I products RNase-free?
Most of our DNase I products are guaranteed free of RNase activity. However, please note that Cat. No. 18047-019 is not tested for RNAse and is recommended primarily for protein applications. The other products are suitable for removing DNA from both RNA and protein preparations, for nick translating DNA, and for generating random fragments of DNA. For more demanding RT-PCR applications, we recommend using DNAse I, Amplification Grade.
What are the ideal storage conditions for the TURBO DNase (2 U/μl) (Cat. No. AM2239, AM2238)? Will the enzyme remain stable even after multiple freeze/thaw cycles?
The TURBO DNAse itself is contained in a buffer containing glycerol, and therefore it will not freeze at -20°C. The reaction buffer is an aqueous salt solution which will remain stable even after multiple freeze/thaw cycles. We do not recommend storing it under different conditions. You could also aliquot the DNAse, but under these circumstances it wouldn't be necessary.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Does TURBO DNase contain DTT?
TURBO DNase does not contain DTT.
Can I purchase the 10X Reaction Buffer included with TURBO DNase as a stand-alone product?
We do not offer the 10X Reaction Buffer that comes with TURBO DNase as a stand-alone item and the composition of the buffer is proprietary.
Does TURBO DNase cleave both single- and double-stranded DNA?
Yes, since the cleavage site for TURBO DNase is the phosphodiester bond, it cleaves both single- and double-stranded DNA.
How does the dsDNase in Maxima First Strand cDNA Synthesis Kit for RT-qPCR, with dsDNase (Cat. Nos. K1671 and K1672) compare with TURBO DNase?
The main difference between the two enzymes is that dsDNase is heat-labile (easily inactivated by moderate heat treatment at 55 degrees C) and specific to dsDNA. Therefore, it does not need a separate heat inactivation step. It gets inactivated during the reverse transcription reaction. Turbo DNase is not heat-labile and is able to degrade both ssDNA and dsDNA. It needs to be heat inactivated post-digestion or has to be removed by phenol-chloroform extraction.
I am getting MagMAX bead carryover in some wells of my plate. What might be causing this and how can I resolve it?
Some bead carryover is common and has not been documented to affect downstream assays. The most common cause of bead carryover is too much input sample. The best way to determine if the sample amount is the cause of the issue is to run PBS with Xeno (artificial DNA/RNA) in the lysis buffer and then check if there is still bead carryover. If that is not the cause, the initial sample may require special processing, such as bead beating or liquification. If there is still bead carryover without the original sample, and in particular, if the carryover is present in the same wells for multiple runs, then this may be an instrument alignment issue.
Do I need any additional consumables when using MagMAX kits on the MagMAX Express-96 instrument or a KingFisher instrument?
Plastic consumables need to be purchased separately in order to use the MagMAX reagents on the MagMAX Express-96 or a KingFisher instrument. The particular consumables and the amount required will vary by kit and protocol so please check the user guide for the particular MagMAX kit to be sure that all required plastic consumables are available prior to sample processing.
Where can I find Bindlt script protocol (.bdz) files for use of MagMAX kits with the KingFisher Flex Purification System and KingFisher Duo Prime Purification System?
The best place to download KingFisher Flex and KingFisher Duo Prime protocols for a particular MagMAX kit is the relevant MagMAX kit product page. The Bindlt script protocol (.bdz) files can be found under the Documents>Product literature section.
Can I scale up the MagMAX kit reaction volumes? If so, how should I do this?
Yes, reaction volumes for MagMAX kits can be scaled up. In general, the amount of MagMAX reagents should be scaled up proportionally for the increase in sample input amount. Please see the specific kit user guide for any recommendation. If none are included, then optimization may be necessary.
Will addition of BME (2-mercaptoethanol) to the MagMAX lysis buffer cause any issues with nucleic acid extraction?
Addition of BME (2-mercaptoethanol) to the lysis buffer of MagMAX kits is fine. This can be beneficial with more RNase-rich samples like certain types of tissues.
In general, we recommend adding 0.3 µL BME per 100 µL of lysate.
Is the TURBO DNase in the MagMAX mirVana Total RNA Isolation Kit (Cat. No. A27828) the same as the standalone TURBO DNase (Cat. No. AM2239)?
The TURBO DNase in the MagMAX mirVana Total RNA Isolation Kit is the same enzyme as the standalone TURBO DNase, but the concentrations of the enzyme differs and therefore it is not an exact equivalent. The TURBO DNase in the MagMAX mirVana Total RNA Isolation Kit is at 20 U/µL, whereas the standalone TURBO DNase is at 2 U/µL.
I want to use the MagMAX-96 for Microarrays Total RNA Isolation Kit to isolate total RNA with the modified spin protocol (https://bit.ly/2Z9cPQK). Can I add the TURBO DNase steps from the standard no-spin protocol to the modified spin protocol?
When using the MagMAX-96 for Microarrays Total RNA Isolation Kit modified spin protocol (https://www.thermofisher.com/us/en/home/references/protocols/nucleic-acid-purification-and-analysis/rna-protocol/isolating-small-rnaas-using-the-magmax-96-for-microarrays-kit.html) for isolating large + small RNA, we recommend going through the RNA isolation procedure and then treating the eluted RNA with TURBO DNase.