UltraPure™ BSA (50 mg/mL)
Invitrogen™

UltraPure™ BSA (50 mg/mL)

UltraPure BSA is a non-acetylated bovine serum albumin (BSA) pure enough to use when DNA or RNA integrity is critical.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM26161 x 50 mg
AM26185 x 50 mg
Catalog number AM2616
Price (TWD)
4,930.00
Online offer
Ends: 31-Dec-2026
6,570.00
Save 1,640.00 (25%)
Each
Quantity:
1 x 50 mg
Price (TWD)
4,930.00
Online offer
Ends: 31-Dec-2026
6,570.00
Save 1,640.00 (25%)
Each
UltraPure BSA is a non-acetylated bovine serum albumin (BSA) pure enough to use when DNA or RNA integrity is critical. One tube containing 50 mg is provided, at a concentration of 50 mg/mL. BSA has many uses as a carrier protein and as a stabilizing agent in enzymatic reactions. In northern, Southern, and dot blot hybridizations, BSA is also used as a blocking agent. It is recommended in buffers for nick translation, polymerase reactions, and ligations. BSA is also a common additive for PCR amplifications, footprinting, and gel shift assays. In restriction digests, BSA has been shown to enhance enzyme activity. The molecular weight is 68 kD. UltraPure BSA is rigorously tested for DNase, RNase, endonuclease, and protease activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAmbion
Quantity1 x 50 mg
Shipping ConditionDry Ice
Concentration50 mg/mL
FormLiquid
Unit SizeEach
Contents & Storage
Store at –20°C.

Frequently asked questions (FAQs)

What is BSA and its function in PCR? What concentration of BSA do you recommend?

BSA, bovine serum albumin, is usually used in PCR as a stabilizer. A concentration of 160 µg/mL is typical. Gelatin may also be used as a stabilizer at 0.01% (w/v).

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.