Yeast RNA (5 mg/mL)
Invitrogen™

Yeast RNA (5 mg/mL)

Purified Saccharomyces cerevisiae yeast RNA is is a very effective blocking agent when used in Northern prehybridization and hybridization buffers, as well as being suitable as a coprecipitant in nucleic acid precipitations.
Have Questions?
Catalog NumberQuantity
AM7120G1 x 0.5 mL Tube
Catalog number AM7120G
Price (USD)
125.65
Online Exclusive
131.00
Save 5.35 (4%)
Each
Quantity:
1 x 0.5 mL Tube
Price (USD)
125.65
Online Exclusive
131.00
Save 5.35 (4%)
Each

Purified Saccharomyces cerevisiae yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant. This total RNA preparation is supplied in one tube containing 0.5 mL at a concentration of 5 mg/mL in diethyl pyrocarbonate-treated distilled water. It is typically used at a working concentration of 10–20 μg/mL.

What is a Coprecipitant?

Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Carrier for Nucleic Acid Precipitation
Product TypeYeast RNA
Quantity1 x 0.5 mL Tube
Shipping ConditionDry Ice
Concentration5 mg/mL
Unit SizeEach
Contents & Storage
Store at –20°C.