Product Image
Invitrogen™
Leukemia (K-562) Total RNA
The highest-quality RNA available from Ambion®, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 100Read more
Have Questions?
Catalog NumberQuantity
AM7832100 μg
Catalog number AM7832
Price (USD)
439.65
Online exclusive
468.00
Save 28.35
Each
Quantity:
100 μg
Price (USD)
439.65
Online exclusive
468.00
Save 28.35
Each
The highest-quality RNA available from Ambion®, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 100 μg is provided at a concentration of 1 mg/mL. Intactness of mRNA, genomic DNA contamination, presence/absence of enzymatic inhibitors, and ribosomal RNA contamination in poly(A) preparations are all important aspects used to judge the quality of FirstChoice® RNAs. The samples are processed using Ambion® RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Note: While it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent® 2100 bioanalyzer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineK-562
Concentration1 mg/mL
Product LineAmbion
Quantity100 μg
Sample SourceCell Line
Sample TreatmentDNase-Treated
Shipping ConditionDry Ice
Sample TypeTotal RNA
SpeciesHuman
Unit SizeEach
Contents & Storage
Store at or below -70°C. Avoid multiple freeze–thaw cycles. Aliquots of the product may be stored short-term at -20°C. DO NOT STORE IN FROST-FREE FREEZER.
Frequently asked questions (FAQs)
Do you offer isolated, purified RNA?
Yes, please visit this website (http://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/purified-rna.html) for our FirstChoice total RNA isolated from human tissues, as well as from E. coli and human cell lines.