UTP Soln. (10 mM)
Invitrogen™

UTP Soln. (10 mM)

This Ambion® ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript® Kits (SKU#s AM1312, AM1316, AM1314, AM1326, AM1324, AM1318,#AM1322,Read more
Have Questions?
Catalog NumberQuantity
AM8140G100 μL
Catalog number AM8140G
Price (USD)
128.00
Each
Quantity:
100 μL
Price (USD)
128.00
Each
This Ambion® ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript® Kits (SKU#s AM1312, AM1316, AM1314, AM1326, AM1324, AM1318,#AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)In Vitro transcription
Label or DyeUnlabeled
Product LineAmbion
Quantity100 μL
Shipping ConditionDry Ice
Concentration10 mM
Unit SizeEach
Contents & Storage
Store at -20°C.