DNase I Buffer (10X)
Invitrogen™

DNase I Buffer (10X)

An Invitrogen component of the MAXIscript™ Kits, the buffer is supplied in one tube containing 1 mL.
Have Questions?
Catalog NumberQuantity
AM8170G1 mL
Catalog number AM8170G
Price (USD)
157.65
Online exclusive
166.00
Save 8.35
Each
Quantity:
1 mL
Price (USD)
157.65
Online exclusive
166.00
Save 8.35
Each

An Invitrogen component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308), the buffer is supplied in one tube containing 1 mL.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Reverse Transcription
Product LineAmbion
Quantity1 mL
Shipping ConditionRoom Temperature
Concentration10X
Unit SizeEach
Contents & Storage
Store at –20°C.