SlideHyb™ Glass Array Hybridization Buffer No. 3 (2 ml tube) - FAQs
SlideHyb™ Glass Array Hybridization Buffer No. 3 (2 ml tube) - FAQs
View additional product information for SlideHyb™ Glass Array Hybridization Buffer No. 3 (2 ml tube) - FAQs (AM8863)
1 product FAQs found
Can Northern blotting be used to detect small RNAs?
Yes. You can use a 15% denaturing polyacrylamide gel for Northern analysis of small RNAs, such as miRNAs and siRNAs. A hyridization buffer optimized for use with short probes, such as ULTRAhyb-Oligo solution, should be used for the best results. For more sensitive detection, enrich the RNA sample for small RNAs (e.g., with the mirVana miRNA Isolation Kit or mirVana PARIS RNA and Native Protein Purification Kit. Of course, using a solution hybridization assay such as the mirVana miRNA Detection Kit to analyze small RNAs can provide much greater sensitivity and allow you to detect multiple small RNAs simultaneously.