IPTG (Dioxane-Free)
Invitrogen™

IPTG (Dioxane-Free)

Ambion IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determineRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM94645 g
AM94622 g
Catalog number AM9464
Price (USD)
283.36
Special offer
Online exclusive
Ends: 27-Jun-2026
322.00
Save 38.64 (12%)
Each
Quantity:
5 g
Price (USD)
283.36
Special offer
Online exclusive
Ends: 27-Jun-2026
322.00
Save 38.64 (12%)
Each

Ambion IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage. IPTG is also used to induce the expression of cloned genes that are under control of the lac operon. Supplied in one bottle containing 5 g.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialIPTG (Isopropyl β-D-thiogalactopyranoside)
Packaging TypeBottle
Product LineAmbion
Quantity5 g
Shipping ConditionRoom Temperature
FormPowder
Unit SizeEach
Contents & Storage
Store at –20°C.