GlycoBlue™ Coprecipitant (15 mg/mL)
Invitrogen™

GlycoBlue™ Coprecipitant (15 mg/mL)

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM95165 x 300 μL
AM9515300 μL
Catalog number AM9516
Price (USD)
315.65
Online Exclusive
336.00
Save 20.35 (6%)
Each
Quantity:
5 x 300 μL
Price (USD)
315.65
Online Exclusive
336.00
Save 20.35 (6%)
Each
GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant. The attached dye increases visibility of the pellet. This product is an ideal coprecipitant in nuclease protection assays at 1/100 dilution of stock solution.

Features of GlycoBlue Coprecipitant:
• Ideal for RT-PCR
• Increases pellet mass and visibility
• Quantitative recovery of low concentrations (ng/mL) of nucleic acid
• Prevents pellet loss in nuclease protection assays

What is a coprecipitant?
Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.

GlycoBlue Coprecipitant can be added to nucleic acid solutions at a final concentration of 50–150 μg/mL. When a typical acetate/alcohol precipitation is done, the GlycoBlue Coprecipitant will precipitate with the nucleic acids, facilitating good RNA or DNA recovery while increasing the size and visibility of the pellet. Since glycogen does not contain appreciable amounts of nucleic acids, it is often preferable to yeast RNA as a coprecipitant, especially in applications where nucleic acid mass is being assessed or where added nucleic acid could interfere or compete with subsequent enzymatic reactions.

The glycogen used is isolated from rabbit liver, a biological source, as are most other preparations of this coprecipitant. Glycogen is treated with Proteinase K and SDS to remove any contaminating nucleases, and then phenol/chloroform extracted, ethanol precipitated, and resuspended in nuclease-free water. The glycogen is guaranteed RNase- and DNase-free.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration15 mg/mL
FormLiquid
Packaging TypeTube
Product LineAmbion, GlycoBlue
PurityMolecular Biology Grade
Quantity5 x 300 μL
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store at –20°C.