Potassium Acetate (3 M), pH 5.5, RNase-free
Invitrogen™

Potassium Acetate (3 M), pH 5.5, RNase-free

Ambion® Molecular biology grade, 3 M Potassium Acetate, pH 5.5 solution is supplied in one bottle containing 100 mL. TheRead more
Have Questions?
Catalog NumberQuantity
AM9610100 mL
Catalog number AM9610
Price (USD)
65.75
Each
Quantity:
100 mL
Price (USD)
65.75
Each
Ambion® Molecular biology grade, 3 M Potassium Acetate, pH 5.5 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Ambion®'s nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPotassium Acetate
For Use With (Application)RNA Applications
Packaging TypeBottle
Product LineAmbion
PurityMolecular Biology Grade
Quantity100 mL
Shipping ConditionRoom Temperature
Concentration3 M
FormLiquid
pH5.5
Unit SizeEach
Contents & Storage
Store at room temperature.