Water-Saturated Phenol, pH 6.6

Catalog number: AM9710

Ambion™  Related applications: DNA Extraction | RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® Water Saturated Phenol is supplied premixed and at pH 6.6 (pH 7.9 ± 0.2, with included buffer). Water Saturated Phenol is used in RNA and DNA extraction procedures and is provided in one bottle of 100 mL. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion® premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name: Phenols
Form: Liquid
Product Line: Ambion™
Product Size: 100 mL
Purity or Quality Grade: Molecular Biology Grade
pH: pH 6.6
Shipping Condition: Room Temperature

Contents & storage

Store at 4°C or –20°C.

Documents

Manuals & protocols

Recommended products