NaCl (5 M), RNase-free
Invitrogen™

NaCl (5 M), RNase-free

Invitrogen Molecular biology grade, 5 M NaCl solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM9760G100 mL
AM9759500 mL
Catalog number AM9760G
Price (USD)
62.65
Online exclusive
66.25
Save 3.60
Each
Quantity:
100 mL
Price (USD)
62.65
Online exclusive
66.25
Save 3.60
Each

Invitrogen Molecular biology grade, 5 M NaCl solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical Name or MaterialSodium Chloride
Packaging TypeBottle
Product LineAmbion
PurityMolecular Biology Grade
Quantity100 mL
Shipping ConditionRoom Temperature
Concentration5 M
FormLiquid
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What can I do to prevent my DNA standards from smearing during electrophoresis?

Add 10 to 20 mM NaCl to the sample to maintain proper ionic strength.