SSC (20X), RNase-free
Invitrogen™

SSC (20X), RNase-free

Molecular biology grade, Invitrogen 20X SSC solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM97631L
AM9770500 mL
AM97654 x 1 L
Catalog number AM9763
Price (USD)
105.65
Online exclusive
111.00
Save 5.35
Each
Quantity:
1L
Price (USD)
105.65
Online exclusive
111.00
Save 5.35
Each

Molecular biology grade, Invitrogen 20X SSC solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen™'s nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Packaging TypeBottle
Product LineAmbion
Quantity1L
Shipping ConditionRoom Temperature
Concentration20X
FormLiquid
Unit SizeEach
Contents & Storage
Store at room temperature.