Tris (1 M), pH 8.0, RNase-free, 100 mL - FAQs
Tris (1 M), pH 8.0, RNase-free, 100 mL - FAQs
View additional product information for Tris (1 M), pH 8.0, RNase-free - FAQs (AM9855G, AM9856)
1 product FAQs found
What alternative do you offer for UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025)?
We recommend Tris (1 M), pH 8.0, RNase‐free (Cat. No. AM9855G).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.