DEPC-Treated Water
Invitrogen™

DEPC-Treated Water

Ambion DEPC-treated water is certified nuclease-free and supplied in five bottles containing 100 mL each. DEPC-treated water is autoclaved pre-Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 x 50 mL
1 x 100 mL
1 x 500 mL
5 x 100 mL
1 x 1000 mL
4 x 1000 mL
Catalog number AM9920
Price (USD)
107.65
Online exclusive
113.00
Save 5.35
Each
Quantity:
1 x 500 mL
Price (USD)
107.65
Online exclusive
113.00
Save 5.35
Each

Ambion DEPC-treated water is certified nuclease-free and supplied in five bottles containing 100 mL each. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Product LineAmbion
Purification MethodAutoclaved, Membrane-Filtered
Recommended StorageStore at room temperature.
Shipping ConditionRoom Temperature
Treatment(s)DEPC-Treated
GradeMolecular Biology
Quantity1 x 500 mL
pH6 to 8
Unit SizeEach