Especificaciones
Tipo de productoFour-Run Expandable I/O Flow Computer
Clasificación de carcasaNEMA 4X
Potencia de entrada+5.5 to +16 VDC
Certificaciones/conformidadUL and c-UL Class I, Div. 2, Groups C and D. UL and c-UL Class I, Div. 1, Group D, Intrinsically safe
Protección de E/SAll I/O lightening/surge protected, meets or exceeds IEEE 472
Intervalo de humedad de funcionamiento0 to 95% RH, Non-condensing
Intervalo de temperatura de funcionamiento-40°C to +85°C (-40°F to +185°F)
Consumo de corriente inactivaMain Board (only): 150µA at 6VDC; 250µA at 12VDC. Main and Expansion Board: 5mA at 6VDC; 5mA at 12VDC
Unit SizeEach