EVOS™ Power Cord, Type I (Australia)
Thermo Scientific™
EVOS™ Power Cord, Type I (Australia)
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries withLeia mais
Have Questions?
Número do catálogoQuantity
AMEP46471 each
Número do catálogo AMEP4647
Preço (BRL)
-
Quantity:
1 each
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type I power plugs. The Type I electrical plug is mainly used in Australia and New Zealand, but is also used in China and parts of Asia, South America, and Africa.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Connector TypesType I (Electrical Plug), Type I (Power Plug)
For Use With (Equipment)EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Quantity1 each
Product LineEVOS
TypePower Cord
Unit SizeEach