EVOS® DVI Video Capture Peripheral
Catalog number: AMEP4717
 Related applications: Cellular Imaging
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The EVOS® DVI Video Capture Peripheral is a portable DVI-to-USB frame grabber ideally suited to capture video output from time-lapse acquisitions in real time. It is compatible with all EVOS® imaging systems that offer DVI output and can capture resolutions up to 1920 x 1200 pixels at a capture rate of up to 30 frames per second.
Explore the entire EVOS® line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): EVOS® FL Auto Imaging System, EVOS® FL Color Imaging System, EVOS® FL Imaging System, EVOS® XL Imaging System
Product Line: EVOS®
Product Size: 1 each
Quantity: 1 each
Type: Other Accessories
Documents
Manuals & protocols
Recommended products