Experience nuclei counting with fluorescent-mode counting algorithms, simplified workflows, built-in dilution calculators and reports using the latest version of Countess software with optional 21 CFR Part 11 compliance. Download now!
Countess™ Light Cube Removal Tool
Invitrogen™
Countess™ Light Cube Removal Tool
Install or remove EVOS Light Cubes in the Countess II FL and Countess 3 FL automated cell counters with the Countess Light Cube Removal Tool. This tool is included with the Countess II FL and 3 FL automated cell counters or a replacement Countess Light Cube Removal Tool can be purchased separately.
Have Questions?
Catalog NumberQuantity
AMEP47471 unit
Catalog number AMEP4747
Price (USD)
44.27
Special offer
Online exclusive
Ends: 27-Jun-2026
46.60
Save 2.33 (5%)
Each
Quantity:
1 unit
Price (USD)
44.27
Special offer
Online exclusive
Ends: 27-Jun-2026
46.60
Save 2.33 (5%)
Each
The Countess Light Cube Removal Tool is for use with the Countess II FL and Countess 3 FL automated cell counters. It is used for installation or removal of EVOS Light Cubes from the instruments. The Countess II FL and Countess 3 FL automated cell counters can accommodate up to two Light Cubes simultaneously. With the Countess Light Cube Removal Tool, users can easily remove and install the EVOS Light Cubes, allowing the flexibility to count cells using any of the 15 available fluorescence channels provided by the Light Cubes.
This tool is included with the Countess II FL and 3 FL automated cell counters and can be conveniently stored in the back panel of the instruments when not in use. A replacement Countess Light Cube Removal Tool (Cat. No. AMEP4747) is offered here for separate purchase.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Cell Counting
For Use With (Equipment)Countess II FL and 3 FL Automated Cell Counters
Quantity1 unit
Shipping ConditionRoom Temperature
Product LineCountess
TypeLight Cube Remove Tool
Unit SizeEach