EVOS™ Onstage Vessel Holder, one Nunc™ multi-well dish - FAQs
EVOS™ Onstage Vessel Holder, one Nunc™ multi-well dish - FAQs
View additional product information for EVOS™ Onstage Vessel Holder, one Nunc™ multi-well dish - FAQs (AMEPVH040)
3 product FAQs found
How should the EVOS vessel holders be stored?
Store the EVOS vessel holders flat and protected from any contact with heavy objects as they can be distorted. Once the holders are distorted and no longer lay flat on the stage, they cannot be used. Store them individually in their own lidded plastic containers or small sturdy boxes.
How should I clean my EVOS vessel holders?
Wipe the vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol or 4,000 ppm hydrogen peroxide (H2O2).
I have an EVOS vessel that does not fit into any of the vessel holders that you offer. What are my options?
We do offer an option for making custom vessel holders. Please contact your Sales Representative for initiating a custom order.