See more signal from immunofluorescence images with Prolong® Diamond. Learn more.

Goat F(ab')2 Anti-Mouse IgM, FITC

Catalog number: AMI4608

Invitrogen™  Related applications: Cellular Imaging | Flow Cytometry | Protein Assays and Analysis | Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Goat F(ab')2 anti-mouse immunoglobulin, human Ig adsorbed, 0.7 mg⁄mL

Format/Formulation: FITC Conjugate

Application: FC
Analyte Specific Reagent. Analytical and performance characteristics are not established.

Specifications

Reactivity: Mouse
Target Isotype: IgM
Host Species: Goat
Antibody Fragment: F(ab')2
Label or Dye: FITC (Fluorescein)
Product Size: 1 mL
Shipping Condition: Dry Ice
Validated Application: Flow Cytometry, Immunofluorescence, Western Blot

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products