Search
View additional product information for Countess™ 3 FL Automated Cell Counter - FAQs (A49866, A50303, AMQAF2000, A49892, AMQAF2001, A50299, A49893)
8 product FAQs found
We suggest the Countess 3 and 3 FL Automated Cell Counters with starter packages; please see the product pages linked below:
Countess 3 FL Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50303)
Countess 3 FL Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50299)
Countess 3 FL Automated Cell Counter Starter Package (Cat. No. A49866)
Countess 3 Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50302)
Countess 3 Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50298)
Countess 3 Automated Cell Counter Starter Package (Cat. No A49865)
For more information about out Countess 3 Automated Cell Counters See the brochure linked below:
linkCountess 3 Automated Cell Counters: A counter that thinks like a cell biologist
The reason for this difference is that the bright field‐only counting assumes that the sample is diluted 1:1 and corrects the result automatically by multiplying it with 2. When you count in the GFP-mode the instrument assumes trypan blue was not used and thus sample not diluted. The total cell concentration displayed after a fluorescent count does not take any dilution into account. Therefore, the results reflect the actual cell concentration in the sample slide, which must be multiplied by any dilution factor present to calculate the original cell concentration.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
The suitable alternative would be Erythrosin B (Cat. No. 409451000, A14180.36, 189460250 and J62619.22).
No. The Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) parameter settings, especially brightness, can vary between instruments due to slight differences in lamp output and other factors.
The Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) FCS files can be uploaded to most flow cytometry software that can accept FCS files, such as FlowJo or the Attune NxT software. Using the flow cytometry software, you may see the distribution of all cells and adjust/fine-tune parameter settings for only the cells of interest or analyze outliers.
With the exception of EVOS Light Cube, White (Cat no. AMEP4672) any of the EVOS Light Cubes for fluorescence detection can be used on the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000).
Yes, with software update version 1.3.1324.552, nuclei can be counted using the Countess 3 FL Automated Cell Counter. We recommend performing FL-based counting. Please refer to this application note for detailed instructions.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
Cells ranging in size from 5 µm to 60 µm and cell suspension density ranging from 1 x 10E5 to 10E6 cells/mL are optimal for counting with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000).