Product Image
Thermo Scientific™
Oxoid™ AnaeroGen™ Compact Sealing Clip
Use with AnaeroGen Compact No. AN0010C; Compact Plastic Pouch, No. AG0020C; Compact Sachet, No. AN0020D; Thermo Scientific™ CampyGen™ Compact Sachet, No. CN0020C.
Have Questions?
Número do catálogoQuantity
AN0005C5/Pk.
Número do catálogo AN0005C
Preço (BRL)
-
Quantity:
5/Pk.
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sealing Clip is for use with AnaeroGen Compact Part No. AN0010C; Compact Plastic Pouch, Part No. AG0020C; AnaeroGen Compact Sachet, Part No. AN0020D; Thermo Scientific™ CampyGen™ Compact Sachet, Part No. CN0020C.
Especificações
Quantity5/Pk.
Product LineThermo Scientific Oxoid
TypeCompact Sealing Clip
Unit SizeEach