Product Image
Thermo Scientific™
Oxoid™ AnaeroGen™ W-Zip Compact Gas Generator System
Oxoid AnaeroGen W-Zip Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
Have Questions?
Catalog NumberQuantity
AN0010W10 Each/Pk.
Catalog number AN0010W
Price (TWD)
-
Quantity:
10 Each/Pk.
Oxoid AnaeroGen W-Zip Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
  • Use 1 sachet / Compact Plastic Pouch
Includes: 10 sachets and 10 W-Zip pouches.
Specifications
Includes10 sachets and 10 W-Zip pouches
Quantity10 Each/Pk.
FormatCompact Gas Generator Kit
Product LineThermo Scientific Oxoid
TypeAnaeroGen W-Zip Compact
Unit SizeEach