Thermo Scientific™
Footswitch
Related applications:
Anatomical Pathology
Operate the Thermo Scientific™ Footswitch with the Thermo Scientific™ HistoStar™ Embedding Workstation, allowing foot operation of the wax tap and leaving both hands free for tissue alignment.
Back to top
Description
Operate the Thermo Scientific™ Footswitch with the Thermo Scientific™ HistoStar™ Embedding Workstation, allowing foot operation of the wax tap and leaving both hands free for tissue alignment.