AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
AquaPro™ Multi-Input Intelligent Process Transmitters
Thermo Scientific™

AquaPro™ Multi-Input Intelligent Process Transmitters

Streamline process monitoring with the Thermo Scientific™ AquaPro™ Multi-Input Intelligent Transmitter. The AquaPro provides accurate, user-friendly monitoring in-line via a reliable, simplified interface, and can be upgraded in the field to accept up to four inputs.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoChannel 1Tensión
AP2XXXKORP100/240 V
AP2XXXJORP24 V
AP3XXXKConductividad de contacto100/240 V
AP3XXXJConductividad de contacto24 V
AP4XXXJConductividad toroidal24 V
AP5XXXKOxígeno disuelto100/240 V
AP5XXXJOxígeno disuelto24 V
AP6XXXKOzono disuelto100/240 V
AP6XXXJOzono disuelto24 V
AP7XXXJDO óptico de tecnología de oxígeno disuelto resistente Pro-X24 V
AP8XXXJAquaSensors DataStick24 V
AP9XXXKpH ultrapuro100/240 V
AP9XXXJpH ultrapuro24 V
AP1XXXJpH24 VCC
AP1XXXKpH90/240 V
AP4XXXKConductividad toroidal90/240 V
AP7XXXKDO óptico de tecnología de oxígeno disuelto resistente Pro-X90/240 V
AP8XXXKAquaSensors DataStick90/240 V
Número de catálogo AP2XXXK
Precio (MXN)
-
Channel 1:
ORP
Tensión:
100/240 V
Streamline your operation with the Thermo Scientific™ AquaPro™ Multi-Input Intelligent Transmitter, with pre-installed sensing card. Featuring the ability to simultaneously display up to four (4) inputs or parameters, the AquaPro provides accurate, user-friendly monitoring of in-line process applications via a compact, simplified interface. From large industrial operations, to small municipal plants, the AquaPro offers the flexibility and versatility required to meet a variety of measurement needs.

Functional design

Obtain a comprehensive overview of your processes by way of a detailed picture on a single display. With the AquaPro you can reduce the time, labor and capital equipment costs associated with having multiple transmitters (installation, operation, maintenance, upgrades, etc.). Engineered to be straight-forward and intuitive, the monitor and user-interface help make your job easier. Containing a large 4.3 in. (109.22 mm) backlit color display, the AquaPro provides crystal-clear visibility in various lighting conditions. Color-coded alerts help communicate errors and warning signals, while operational menus remain simple and direct.

Unmatched versatility

Whether in the field, industrial plant, or on the manufacturing floor, the AquaPro keeps you primed and prepped to handle a variety of situations. Operators have the ability to monitor a broad range of parameters for diverse applications. Equipped with one (1) sensor card as standard, users can track up to four (4) sensors (analog or digital) by simply adding additional plug-and-play cards. Monitor pH/ORP, conductivity, dissolved oxygen, dissolved ozone, turbidity and suspended solids- the AquaPro is up for the challenge.

Purpose built

Designed to tackle the realities and utility of industrial operations, the AquaPro is built to last. Housed in a UV-resistant polycarbonate case, with NEMA 4X and IP66 waterproof ratings, you can trust that the AquaPro will remain operable in a variety of harsh industrial environments. Adaptable mounting options also provide the ability to configure the transmitter to your needs- the AquaPro is easily pipe, panel or wall-mounted.

Clear communication

Supporting most major communication protocols, the AquaPro can be used with a wide range of data systems for different applications- including information transfer to PLC, DCS, SCADA and other PC-based control systems and 0/4-20mA, MODBUS™ RTU, Ethernet/IP and Modbus TCP. Using the USB data port, data can be easily downloaded and new firmware quickly installed. This means new features and capabilities without stopping to change hardware. And in instances where additional transmitters are required, the configuration of an existing AquaPro can be easily saved and copied to a newly installed AquaPro using a removable USB drive.

Especificaciones
Salidas analógicasTwo isolated 0/4-20mA per channel (non-reversable), 0.01mA resolution, linear/log/PID capability
Material del envasePolycarbonate
Channel 1ORP
Canales4 (Customizable with modules)
Detalles de comunicaciónOptional MODBUS RTU or Ethernet/IP
DIN Size1/2 DIN
DescripciónIntelligent multi-input transmitter with pre-installed sensing card for analog ORP sensors
Dimensiones (Largo x Ancho x Alto)144 x 144 x 128 mm (5.67 x 5.67 x 5.01 in.)
Tipo de pantallaColor Graphic LCD
Requisitos eléctricos100 to 240 V, 9.6 W, 50/60 Hz
Clasificación de carcasaNEMA 4X, IP66
Para utilizar con (equipo)Aquasensors AnalogPlus ORP sensors
Hazardous LocationsClass 1 Div 2, Groups A, B, C, D (in combination with approved sensors)
Incluye109.22 mm (4.3 in.) high-contrast color graphical display
Tipo de tecladoTactile keypad with 7-key menu navigation
Humedad relativa máx.95%, non-condensing
Opciones de montajeWall, panel or pipe
Detalles del reléThree form C relays, 5A @ 250VAC
Temperatura de funcionamiento (imperial)-4°F to +140°F
Interfaz USBFront-panel accessible USB-A for firmware updates and datalog downloads
Tensión100/240 V
Peso (imperial)1.3 kg
Peso (métrico)3 lb.
Temperatura de funcionamiento (métrico)-20°C to +60°C
Unit SizeEach