Thermo Scientific™
AP2/AP5 Series Alpha Scintillation Probes
 Related applications:
Radiation Detection Measurement
Conclusively locate alpha contamination on personnel or workplace surfaces. Thermo Scientific™ AP2 and AP5 Series Alpha Scintillation Probes are general purpose instruments containing a zinc sulfide alpha scintillator, the preferred technology for detecting alpha contamination on the hands, feet or other surfaces. The AP2 probe has a conveniently angled radiation window and is contained in a light alloy housing for easy self monitoring. The AP5 probe features rugged construction and field-replaceable radiation windows for reliable operation while monitoring workplaces, benches, personnel and tools.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
AP2 Series
50cm2 square window.
The low weight and durability of the probes make them easy to handle.
Conveniently angled radiation window for self monitoring of clothing.
Field-replaceable radiation windows.
Available versions:
AP2/4A with connector type PET
AP2/4AR with connector type PET, ruggedized version
AP2/4B with connector type MHV
AP5 Series
100cm2 rectangular window.
High transmission rectangular hex-grille.
Slim, lightweight, wedge-shaped profile.
Field-replaceable radiation windows.
Available versions:
AP5/AD with connector type PET.
AP5/RA with connector type PET, ruggedized version.
AP5/BD with connector type MHV.
AP5/RB with connector type MHV, ruggedized version.
Information on calibration sources can be provided on request.
Documents
Manuals & protocols