This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

Pathatrix™ 5 Pooling Dual E. coli/Salmonella Kit

Catalog number:  APDES250P

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Pathatrix® 5 Pooling DUAL E. coli/Salmonella Kit, when used with the Pathatrix® Auto Instrument, facilitates the concentration and isolation of E. coli O157:H7 and Salmonella spp. bacteria from pre-enriched food and environmental samples for downstream detection by PCR, immunoassay or culture. This kit format supports pooling of 5 enriched samples into a single test, reducing downstream testing costs by up to 80%.

When used with the Pathatrix® Auto Instrument, the Pathatrix® 5 Pooling DUAL E. coli/Salmonella Kit:
• Helps save money – supports pooling protocols that save up to 80% on downstream detection costs
• Minimizes false positive and indeterminate rates – isolates specific bacteria from background flora and PCR inhibitors
• Reduces time-to-results – concentrates target organisms, so less enrichment time is required

Each kit includes one vial of antibody-coated beads and sufficient numbers of sterile, single-use plastic consumables for 50 pooled tests (250 individual, enriched samples).
For Testing of Food and Environmental Samples Only.

Specifications

Final Product: Bacteria (Intact)
For Use With (Equipment): Pathatrix® Auto System
High Throughput Compatibility: Automated Protocols, High Throughput-Compatible
Isolation Technology: Immunomagnetic Separation
Number of Reactions: 50 Tests
Product Line: Pathatrix™
Product Size: 50 tests
Sample Source: Pooled Samples
Sample Treatment: Overnight Incubation (Pre-Enrichment)
Sample Type (Specific): Environmental Sample, Food
Sample Volume: 10 mL (5 x 10 mL per pool)
Strain or Genotype: E. coli O157:H7, Salmonella spp.
Target Organism Class: E. coli, Salmonella spp.

Contents & storage

Each kit includes one vial of antibody-coated beads and sufficient sterile, single-use plastic consumables for 50 pooled tests (250 individual, enriched samples).

Store beads at 2–8°C; do not freeze.

Documents

Manuals & protocols