AquaTrace™ ppb Dissolved Oxygen System
Actual product may vary
Thermo Scientific™

AquaTrace™ ppb Dissolved Oxygen System

Monitor trace oxygen levels in critical processes to allow quick response and minimize costly downtime with this system.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호전압
AQD1C0100/240 VDC
AQD1A024 VDC
카탈로그 번호 AQD1C0
제품 가격(KRW)
-
전압:
100/240 VDC

Monitor trace oxygen levels in critical processes to allow quick response and minimize costly downtime with the Thermo Scientific™ AquaTrace™ ppb Dissolved Oxygen System, a trace dissolved oxygen sensor that measures accurately in pure water processes. Best performance is achieved when used in applications where process temperature, flow and pressure are stable.

Features:

  • Reagent-free amperometric design
  • Trace DO system includes Trace DO Thermo Scientific™ DataStick™, flow cell, AV38 local display; assembled on the panel for simple installation
  • Temperature compensated
  • Pre-calibrated, plug & play sensors
  • Rugged membrane in replaceable sensor cap
  • Remote measurement, calibration, configuration and diagnostics
  • Convenient turn-key AquaTrace DO monitoring system for reliable low DO measurements with a small footprint

Markets/Applications:

  • Process water, production and distribution: Collect quality control data
  • Food and beverage: Monitor process water quality
  • Pure water analysis: Accurate trace DO measurements and minimal drift

DataStick:

  • Provides universal conversion of sensor signals and interactive communications for measurement, calibration, configuration and diagnostics

Communications Adapter:

  • Plugs into the DataStick to provide power and direct interactive communications with control systems

AV38 Local Display:

  • Two-line display and seven-key navigation
  • Data reporting with up to two current outputs
  • Two Form C relays
  • Digital communications
  • AquaTrace ppb DO System with Trace DO ppb DataStick and flow cell installed
  • Trace ppb Dissolved Oxygen Sensor Head Pre-calibrated for ppb level dissolved oxygen and temperature
  • Can be plugged into any DataStick to yield accurate 24-bit data

AquaTrace ppb DO System:

  • Trace DO ppb DataStick and flow cell installed
Trace ppb Dissolved Oxygen Sensor Head:
  • Precalibrated for ppb level dissolved oxygen and temperature
  • Can be plugged into any DataStick to yield accurate 24-bit data
사양
통신 세부 정보Modbus R TU, DeviceNet, CANopen, EtherNet, USB, RJ-45, RS485
설명ntegral AV38 with two current outputs, two relays, flow chamber
장착 옵션1 in. NPT threads at back of sensor, 1 in. NPT threads at front of sensor
출력1
규제 세부 정보RoHS compliant assembly
릴레이 세부 정보2
온도(영국식 단위) 작동41°F to 122°F
전압100/240 VDC
작동 온도(미터법) 작동5°C to 50°C
유형Dissolved Oxygen System
Unit SizeEach