Product Image
Thermo Scientific™
AquaSensors™ AquaClear Turbidity Measurement System
AquaClear Low range Turbidimeter with AV38 local display 0 to 200 NTU, 1 x 4-20mA, Modbus RTU over RS485 host communication
Have Questions?
Número de catálogoTipo de pantalla
AQT1C4LCD
Número de catálogo AQT1C4
Precio (MXN)
-
Tipo de pantalla:
LCD
AquaClear Low range Turbidimeter with AV38 local display 0 to 200 NTU, 1 x 4-20mA, Modbus RTU over RS485 host communication
Thermo Scientific AquaSensors AquaClear Turbidity Measurement System
The Thermo Scientific AquaSensors AquaClear low-range turbidimeter system delivers accurate turbidity measurements in drinking water applications and other process applications to monitor water quality. The AquaClear turbidimeter uses a pre-calibrated plug-in optical sensor head. The sensor is inserted into a sample chamber specifically designed to condition the water for turbidity measurements. The measurement chamber removes bubbles from the water so that solid
particles can be accurately detected. Maintenance costs are low with the AquaClear system. The lamp provides collimated white light for a minimum of three years and is easily changed. The volume of the measurement chamber is 135 mL and requires a small amount of formazan to perform EPA-mandated calibrations, thus making the AquaClear turbidimeter a cost-effective investment.
The AquaClear system is offered in three convenient versions: a digitally networked version without a local display that can interface digitally with industrial control systems; a digitally networked version with a local display (AV38); and an analog version with a local display (AV88) that cannot digitally communicate to a host system but provides two analog outputs, two relays, and AC/DC power options. Both AV38 and AV88 local displays offer standard commands for calibration, configuration, diagnostics, and troubleshooting
Especificaciones
Métodos aprobados por la EPA180.1
Clasificación IPNot Stated
PrecisiónAccuracy: ±2 % of reading or ±0.015 NTUwhichever is greater. ±5 % of reading above 40 NTU
Resolución0.001 NTU
Garantía12 Months
Selección del intervalo automáticoNo
Tipo de pantallaLCD
Fuente de luzWhite Light
TipoLow Range Turbidity Meter
Unit SizeEach