Interferon gamma Antibody

Catalog number: ARC4833

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

IFN gamma, also called Type II interferon, macrophage activation factor, and immune interferon is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE it appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific and includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFNg is involved in cytokine regulation and also acts synergistically with other cytokines.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Rat
Validated Application: Neutralizing, Immunohistochemistry
Host Species: Rabbit
Clonality: Polyclonal
Target Gene: IFN-gamma
Gene ID: 25712
Label or Dye: Unconjugated
Product Size: 1 mg
Protein Family: Cytokines & Receptors
Research Category: Immunology, Signal Transduction, Virology
Purity or Quality Grade: Azide Free
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products