Assay Ready Plate Production
Assay Ready Plate Production
Assay Ready Plate Production
Assay Ready Plate Production
Assay Ready Plate Production
Thermo Scientific™

Assay Ready Plate Production

Optimize performance, reliability and flexibility using Thermo Scientific™ Assay Ready Plate Production.
Have Questions?
Catalog NumberFor Use With (Application)
ARPP200Assay Plate Production
Catalog number ARPP200
Price (BRL)
-
For Use With (Application):
Assay Plate Production

Optimize performance, reliability and flexibility using Thermo Scientific™ Assay Ready Plate Production. Thermo Scientific automated solutions scale with your operations, from low capacity single assay workstations to full scale production of multiple uHTS assay plate types. Prepare all your assay plates when you need them.

Systems are designed to meet your current capacity challenges and can adapt as assay demands change.
  • Offers the fastest distributed motion platforms designed to address demand for multiple assays simultaneously
  • Systems support all plate and tube racks with a microtitre plate footprint in all well formats (96, 384, 1536)
  • Supports all common Barcode formats
  • Assay plate preparation automation can be configured with your choice of liquid handling, dispensing, centrifugation, capping/decapping, tip handling, incubation, shaking, sealing, piercing, barcode labeling, and storage
  • Optional environmental & safety enclosures to protect samples and personnel
  • All systems run with Momentum™ scheduling and control software for optimal performance, reliability and flexibility
Specifications
DescriptionAssay Ready Plate Production
For Use With (Application)Assay Plate Production
Product TypeAutomated Assay Plate Production
Unit SizeEach