GlowQuant™ ATP Cell Viability Assay Kit
GlowQuant™ ATP Cell Viability Assay Kit
GlowQuant™ ATP Cell Viability Assay Kit
Invitrogen™
GlowQuant™ ATP Cell Viability Assay Kit
The GlowQuant™ ATP Cell Viability Assay Kit is a ready-to-use, bioluminescence-based solution for rapid and sensitive measurement of viable cells through ATP detection and quantification.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
ATP-CV1100100 mL
ATP-CV101010 mL
카탈로그 번호 ATP-CV1100
제품 가격(KRW)
-
수량:
100 mL
The GlowQuant™ ATP Cell Viability Assay Kit is a ready-to-use, bioluminescence-based solution for rapid and sensitive measurement of viable cells through ATP detection and quantification. Designed for wide variety of applications, this luminescence-based ATP assay supports cell health, proliferation studies, and cytotoxicity analysis with a simple one-step workflow.
The assay uses firefly luciferase to catalyze the oxidation of luciferin in the presence of ATP and oxygen, generating a stable bioluminescent signal directly proportional to ATP concentration, and therefore to the number of viable cells.
By measuring intracellular ATP (a universal marker of metabolically active cells), this assay provides a direct and quantitative assessment of cell health.
Key Features:
Features of GlowQuant™ ATP Cell Viability Assay Kit include:
  • High sensitivity ATP detection— Detects low cell numbers, down to ~50 cells/well in a 96-well plate.
  • Fast and simple — One-step add-and-read format with minimal hands-on time and results in as little as 10 minutes
  • No reagent preparation required — Supplied as a ready-to-use, pre-formulated reagent.
  • Stable signal — Bioluminescent glow with a signal half-life > 2 hours for flexible plate reading.
  • Quantitative and reliable performance — Linear response over a wide dynamic range for reliable cell-number estimation.
  • Microplate reader compatible — Designed for use with standard luminescence microplate readers in 96- and 384-well formats; compatible with automation.
How it Works
The GlowQuant™ assay features a simple add-and-read protocol with no separate lysis step required.
  1. Add an equal volume of reagent directly to cultured cells.
  2. Mix for 1 minute.
  3. Incubate at room temperature for 10–20 minutes.
  4. Measure luminescence using a luminometer or compatible microplate reader.
The emitted luminescent signal is proportional to intracellular ATP levels. Because viable cells maintain ATP while damaged or non-viable cells rapidly lose ATP, signal intensity directly reflects the number of metabolically active cells in each well.
Supporting Your Cell Health Research Across Detection Modes
Thermo Fisher Scientific supports cell health research with a broad portfolio of validated assays spanning luminescence, fluorescence, and absorbance detection modes. From rapid ATP-based viability assays to multiplex-compatible fluorescent and colorimetric solutions, our cell health tools are designed to integrate seamlessly with standard plate readers, automated workflows, and high-throughput screening platforms— to help provide scalable solutions backed by global technical support.
For Research Use Only.
사양
세포 유형Adherent and Suspension Cells
검출 방법Luminescence
형태Liquid
형식96-well Plate, 384-well Plate
인큐베이션 시간10 to 20 min.
수량100 mL
연구 카테고리Cell Health
용도(애플리케이션)Cell Viability
용도 (장비)Luminometer, Microplate Reader
제품라인GlowQuant
제품 유형Cell Viability Reagent
Unit SizeEach
구성 및 보관
Store at -20°C, protected from light